Please login to your online account to display your discounted pricing

anti-GABARAP Polyclonal, Thermo Scientific™

GABARAP Polyclonal Ab PA1-18289 from Thermo Scientific™ specifically detects GABARAP in Hu and Rt samples, and it is validated for IF, IHC and WB

$506.05 - $506.05

Specifications

Antigen GABARAP
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-542

 
pierce biotechnology inc
PA118289
Unlabeled 100μL Each for $506.05
Description & Specifications

Specifications

Antigen GABARAP
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminal residues of human GABARAP.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

GABARAP Polyclonal Ab PA1-18289 from Thermo Scientific™ specifically detects GABARAP in Hu and Rt samples, and it is validated for IF, IHC and WB

Gamma-aminobutyric acid A receptors [GABA(A) receptors] are ligand-gated chloride channels that mediate inhibitory neurotransmission. This gene encodes GABA(A) receptor-associated protein, which is highly positively charged in its N-terminus and shares sequence similarity with light chain-3 of microtubule-associated proteins 1A and 1B. This protein clusters neurotransmitter receptors by mediating interaction with the cytoskeleton.
Host Species: Rabbit
Cross Reactivity: Human, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to the N-terminal residues of human GABARAP.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Gamma-aminobutyric acid receptor-associated protein, GABA(A) receptor-associated protein MM46, GABARAP, FLC3B
Uniprot Accession: O95166, P60517
Gene Symbol: GABARAP
Entrez GeneID: 11337, 58974
PA1-18289 detects GABARAP in Human and Rat samples. PA1-18289 has been successfully used in Immunofluorescence, Immunohistochemistry and Western Blot procedures. PA1-18289 The immunogen is Synthetic peptide corresponding to the N-terminal residues of human GABARAP..