missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GABARAP, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region of the human GABARAP protein (within residues 1-50). [Swiss-Prot O95166]
Antigen GABARAP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP197416 Novus Biologicals Canada
NBP197416
0.1mL Each for $309.05
NBP197416SS Novus Biologicals Canada
NBP197416SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
GABARAP
Affinity Purified
RUO
11337
1.12mg/mL
A synthetic peptide made to an N-terminal region of the human GABARAP protein (within residues 1-50). [Swiss-Prot O95166]
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin)
PBS, 30% glycerol