Please login to your online account to display your discounted pricing

anti-GABARAP, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the human GABARAP protein (within residues 50-117). [Swiss-Prot O95166]
Antigen GABARAP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171771SS

 
novus biologicals canada
NBP171771SS
Unlabeled 0.025mL Each for $109.00

NBP171771

 
novus biologicals canada
NBP171771
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the human GABARAP protein (within residues 50-117). [Swiss-Prot O95166]
Antigen GABARAP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human and mouse. Predicted to react with rat based on 100% sequence homology.
Gene Accession No. O95166
Gene ID (Entrez) 11337
Formulation PBS, 30% glycerol
Concentration 1.11mg/mL