missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-G6b/C6orf25, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 170-220 of human G6B was used as the immunogen.
Antigen G6b/C6orf25
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224647SS Novus Biologicals Canada
NBP224647SS
0.025mg Each for $109.00
NBP224647 Novus Biologicals Canada
NBP224647
0.1mg Each for $257.41
Specifications Description & Specifications

Specifications

Rabbit
G6b/C6orf25
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_f), rhesus monkey and chimpanzee, and 72% homologous in cow.
100μg in 200μL PBS containing 0.05% BSA
A portion of amino acids 170-220 of human G6B was used as the immunogen.
Polyclonal
Primary
Western Blot
80739
0.5mg/mL