Please login to your online account to display your discounted pricing

anti-G6b/C6orf25, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human G6B was used as the immunogen.
Antigen G6b/C6orf25
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224505SS

 
novus biologicals canada
NBP224505SS
Unlabeled 0.025mg Each for $109.00

NBP224505

 
novus biologicals canada
NBP224505
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human G6B was used as the immunogen.
Antigen G6b/C6orf25
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b), rhesus monkey, bovine, chicken, horse and pig, 92% homologous in cat, mouse and rat, and 78% homologous in dog.
Gene ID (Entrez) 80739
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL