Please login to your online account to display your discounted pricing

anti-G6b/C6orf25, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224647SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224647SS

  • / Each
 

Description & Specifications

Specifications

Antigen G6b/C6orf25
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 170-220 of human G6B was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 80739
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_f), rhesus monkey and chimpanzee, and 72% homologous in cow.
Target Species Human
Target Species Primate