Please login to your online account to display your discounted pricing

anti-FXR/NR1H4, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of the human FXR protein. [UniProt No. Q96RI1]
Antigen FXR/NR1H4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB400153

 
novus biologicals canada
NB400153
Unlabeled 0.1mL Each for $309.05

NB400153SS

 
novus biologicals canada
NB400153SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of the human FXR protein. [UniProt No. Q96RI1]
Antigen FXR/NR1H4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene ID (Entrez) 9971
Formulation Tris-glycine, 150mM NaCl
Concentration 5.7mg/mL