Please login to your online account to display your discounted pricing

anti-FUS, Polyclonal, Novus Biologicals™

$406.15 - $406.15

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human FUS protein (within residues 1-150). [Swiss-Prot P35637]
Antigen FUS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP150623

 
novus biologicals canada
NBP150623
Unlabeled 0.1mL Each for $406.15
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human FUS protein (within residues 1-150). [Swiss-Prot P35637]
Antigen FUS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and Mouse. Immunogen sequence has 91% identity to bovine.
Gene Accession No. P35637
Gene ID (Entrez) 2521
Formulation PBS, 0.1% BSA, 30% glycerol
Concentration 1.3mg/mL