Please login to your online account to display your discounted pricing

anti-FUS, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP150623SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP150623SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P35637
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 2521
Concentration 1.3mg/mL
Conjugate Unlabeled
Formulation PBS, 0.1% BSA, 30% glycerol
Regulatory Status RUO
Test Specificity Human and Mouse. Immunogen sequence has 91% identity to bovine.
Cross Reactivity Human
Cross Reactivity Mouse
Antigen FUS
Quantity 0.025mL
Immunogen A genomic peptide made to an N-terminal region of the human FUS protein (within residues 1-150). [Swiss-Prot P35637]
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Format Affinity Purified