missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Furin, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 740-790 of human Furin was used as the immunogen.
Antigen Furin
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224687 Novus Biologicals Canada
NBP224687
0.1mg Each for $257.41
NBP224687SS Novus Biologicals Canada
NBP224687SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Furin
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and xenopus and 95% homologous in cow, mouse, rat, chicken and 89% homologous in dog.
5045
0.5mg/mL
A portion of amino acids 740-790 of human Furin was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin)
P09958
100μg in 200μL PBS containing 0.05% BSA