Please login to your online account to display your discounted pricing

Frizzled-6 Rat anti-Mouse, Clone: 211435, R&D Systems

Rat Monoclonal Antibody

$207.26 - $441.90

Specifications

Antigen Frizzled-6
Clone 211435
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB1526050

 
r & d systems
RDSMAB1526050
Unlabeled 50μg Each for $207.26

MAB1526500

 
r & d systems
RDSMAB1526500
Unlabeled 500μg Each for $441.90
Description & Specifications

Specifications

Antigen Frizzled-6
Clone 211435
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-6. His19-Val153 (predicted)
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Frizzled-6 Monoclonal antibody MAB1526 Specifically detects Frizzled-6 in Mouse samples. It is validated for Western Blotting