Please login to your online account to display your discounted pricing

Frizzled-3 Rat anti-Human, Mouse, Clone: 169310, R&D Systems

Rat Monoclonal Antibody

$202.89 - $326.67

Specifications

Antigen Frizzled-3
Clone 169310
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB1001100

 
r & d systems
RDSMAB1001100
Unlabeled 100μg Each for $326.67

MAB1001050

 
r & d systems
RDSMAB1001050
Unlabeled 50μg Each for $202.89
Description & Specifications

Specifications

Antigen Frizzled-3
Clone 169310
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3. Met1-Val157
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Frizzled-3 Monoclonal antibody MAB1 Specifically detects Frizzled-3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry