Please login to your online account to display your discounted pricing

anti-Frizzled-10, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 150-200 of human Frizzled-10 was used as the immunogen
Antigen Frizzled-10
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223659SS

 
novus biologicals canada
NBP223659SS
Unlabeled 0.025mL Each for $139.00

NBP223659

 
novus biologicals canada
NBP223659
Unlabeled 0.1mL Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 150-200 of human Frizzled-10 was used as the immunogen
Antigen Frizzled-10
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse and monkey and 88% homologous in rat.
Gene ID (Entrez) 11211
Formulation 100μL of crude rabbit sera with 50% glycerol