Please login to your online account to display your discounted pricing

Frizzled-1 Rat anti-Human, Mouse, Clone: 162531, R&D Systems

Rat Monoclonal Antibody

$207.26 - $333.71

Specifications

Antigen Frizzled-1
Clone 162531
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB11201050

 
r & d systems
RDSMAB11201050
Unlabeled 50μg Each for $207.26

MAB11201100

 
r & d systems
RDSMAB11201100
Unlabeled 100μg Each for $333.71
Description & Specifications

Specifications

Antigen Frizzled-1
Clone 162531
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled‑1. Gln72-His248 (Met122Ile)
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Frizzled-1 Monoclonal antibody MAB11201 Specifically detects Frizzled-1 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blotting