Please login to your online account to display your discounted pricing

anti-FPRL1/FPR2, Polyclonal, Novus Biologicals™

$214.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to the 2nd extracellular loop of the human FPRL1 protein (between residues 300-350) [UniProt P25090]
Antigen FPRL1/FPR2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NLS1878SS

 
novus biologicals canada
NLS1878SS
Unlabeled 0.025mL Each for $214.00

NLS1878

 
novus biologicals canada
NLS1878
Unlabeled 0.05mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to the 2nd extracellular loop of the human FPRL1 protein (between residues 300-350) [UniProt P25090]
Antigen FPRL1/FPR2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and mouse. Rat reactivity reported in scientific literature (PMID: 24086560).
Gene Accession No. P25090
Gene ID (Entrez) 2358
Formulation PBS, 30% glycerol
Concentration 1.1mg/mL