Please login to your online account to display your discounted pricing

anti-FPR1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 176-190 of human FPR1 (CTFNFSPWTNDPKER) was used as the immunogen
Antigen FPR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056473T

 
novus biologicals canada
NB10056473SS
Unlabeled 0.025mg Each for $139.00

NB10056473

 
novus biologicals canada
NB10056473
Unlabeled 0.1mg Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 176-190 of human FPR1 (CTFNFSPWTNDPKER) was used as the immunogen
Antigen FPR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimp, gorilla and orangutan, and 67% homologous in mouse. Reacts with Chimpanzee, Gorilla, Human and Orangutan. 67% sequence identity with mouse protein.
Gene Accession No. P21462
Gene ID (Entrez) 2357
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL