Please login to your online account to display your discounted pricing

anti-FoxP3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224397SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224397SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_c, and CRA_d), opossum, chimpanzee, rhesus monkey, horse, dog, mouse, cow and rat, 78% homologous in zebrafish and 71% homologous in chicken. In human, the four isoforms range in size from 941 amino acids to 1008 amino acids, with theoretical molecular weights of approximately 103kD to 110kD.
Conjugate Unlabeled
Gene Accession No. Q9BZS1
Gene ID (Entrez) 50943
Primary or Secondary Primary
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Immunogen A synthetic peptide corresponding to the exon 2 deletion site was used as immunogen.
Antigen FoxP3
Regulatory Status RUO
Isotype IgG
Format Purified
Host Species Rabbit
Quantity 0.025mg