Please login to your online account to display your discounted pricing

anti-FoxP3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224397

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224397

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_c, and CRA_d), opossum, chimpanzee, rhesus monkey, horse, dog, mouse, cow and rat, 78% homologous in zebrafish and 71% homologous in chicken. In human, the four isoforms range in size from 941 amino acids to 1008 amino acids, with theoretical molecular weights of approximately 103kD to 110kD.
Gene ID (Entrez) 50943
Concentration 0.5mg/mL
Conjugate Unlabeled
Gene Accession No. Q9BZS1
Regulatory Status RUO
Format Purified
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Isotype IgG
Host Species Rabbit
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Quantity 0.1mg
Antigen FoxP3
Immunogen A synthetic peptide corresponding to the exon 2 deletion site was used as immunogen.