Please login to your online account to display your discounted pricing

anti-FoxP3, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal region of human FOXP3 (between residues 1-50). [UniProt No. Q9BZS1]
Antigen FoxP3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600246

 
novus biologicals canada
NB600246
Unlabeled 0.1mL Each for $309.05

NB600246SS

 
novus biologicals canada
NB600246SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal region of human FOXP3 (between residues 1-50). [UniProt No. Q9BZS1]
Antigen FoxP3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q9BZS1
Gene ID (Entrez) 50943
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL