Please login to your online account to display your discounted pricing

anti-FoxP3, HRP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600246H

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600246H

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene ID (Entrez) 50943
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Conjugate HRP
Format Affinity Purified
Gene Accession No. Q9BZS1
Immunogen A synthetic peptide made to the N-terminal region of human FOXP3 (between residues 1-50). [UniProt No. Q9BZS1]
Host Species Rabbit
Antigen FoxP3
Quantity 0.1mL
Applications Western Blotting
Regulatory Status RUO