Please login to your online account to display your discounted pricing

anti-FoxP3, DyLight 550, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10039002R

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10039002R

  • / Each
 

Description & Specifications

Specifications

Conjugate DyLight 550
Regulatory Status RUO
Antigen FoxP3
Format Purified
Primary or Secondary Primary
Gene ID (Entrez) 50943
Test Specificity Mouse. This antibody shows reactivity against mouse CD4+CD25+T cells. There appears to be no reactivity against human Jurkat and mouse CD4+CD25-T cell lysates. Human reactivity reported in scientific literature (PMID: 23922187).
Formulation 50mM sodium borate
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Applications Flow Cytometry
Applications Western Blotting
Immunogen A peptide corresponding to amino acids 43-100 of mouse FOXP3. [Swiss-Prot No. Q99JB6]
Quantity 0.1mL
Host Species Rabbit