Please login to your online account to display your discounted pricing

anti-FoxP3, DyLight 550, Polyclonal, Novus Biologicals™

$406.00 - $406.00

Specifications

Host Species Rabbit
Immunogen A peptide corresponding to amino acids 43-100 of mouse FOXP3. [Swiss-Prot No. Q99JB6]
Antigen FoxP3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10039002R

 
novus biologicals canada
NB10039002R
DyLight 550 0.1mL Each for $406.00

NB600246R

 
novus biologicals canada
NB600246R
DyLight 550 0.1mL Each for $406.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A peptide corresponding to amino acids 43-100 of mouse FOXP3. [Swiss-Prot No. Q99JB6]
Antigen FoxP3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Test Specificity Mouse. This antibody shows reactivity against mouse CD4+CD25+T cells. There appears to be no reactivity against human Jurkat and mouse CD4+CD25-T cell lysates. Human reactivity reported in scientific literature (PMID: 23922187).
Gene ID (Entrez) 50943
Formulation 50mM sodium borate