Please login to your online account to display your discounted pricing

anti-FOXO3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224579

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224579

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Quantity 0.1mg
Gene ID (Entrez) 2309
Concentration 0.5mg/mL
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. O43524
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee, horse, boar and rhesus monkey, 93% homologous in dog and 87% homologous in cow, mouse and rat.
Format Affinity Purified
Antigen FOXO3
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Immunogen A portion of amino acids 600-673 of human FOXO3A was used as the immunogen.