Please login to your online account to display your discounted pricing

anti-FOXO3, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 600-673 of human FOXO3A was used as the immunogen.
Antigen FOXO3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224579SS

 
novus biologicals canada
NBP224579SS
Unlabeled 0.025mg Each for $109.00

NBP224579

 
novus biologicals canada
NBP224579
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 600-673 of human FOXO3A was used as the immunogen.
Antigen FOXO3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee, horse, boar and rhesus monkey, 93% homologous in dog and 87% homologous in cow, mouse and rat.
Gene Accession No. O43524
Gene ID (Entrez) 2309
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL