Please login to your online account to display your discounted pricing

anti-FoxN1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223665SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223665SS

  • / Each
 

Description & Specifications

Specifications

Antigen FoxN1
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Mouse
Format Antisera
Formulation 100μL of crude rabbit sera with 50% glycerol
Host Species Rabbit
Immunogen A portion of amino acid 450-500 of mouse FOXN1/Hfh11 was used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8456
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and rat, 86% homologous in dog, 73% homologous in cow and 66% homologous in human.
Target Species Murine