Please login to your online account to display your discounted pricing

anti-FoxN1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223665

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223665

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene ID (Entrez) 8456
Primary or Secondary Primary
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and rat, 86% homologous in dog, 73% homologous in cow and 66% homologous in human.
Quantity 0.1mL
Format Antiserum
Applications Western Blotting
Host Species Rabbit
Immunogen A portion of amino acid 450-500 of mouse FOXN1/Hfh11 was used as the immunogen.
Antigen FoxN1
Cross Reactivity Mouse
Regulatory Status RUO