missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-FoxN1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 450-500 of mouse FOXN1/Hfh11 was used as the immunogen.
Antigen FoxN1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP223665 Novus Biologicals Canada
NBP223665
0.1mL Each for $255.80
NBP223665SS Novus Biologicals Canada
NBP223665SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
FoxN1
Antisera
RUO
The amino acid sequence used as immunogen is 100% homologous in mouse and rat, 86% homologous in dog, 73% homologous in cow and 66% homologous in human.
100μL of crude rabbit sera with 50% glycerol
A portion of amino acid 450-500 of mouse FOXN1/Hfh11 was used as the immunogen.
Polyclonal
Primary
Western Blot
8456