Please login to your online account to display your discounted pricing

anti-FOXL2, Polyclonal, Novus Biologicals™

$142.00 - $406.00

Specifications

Host Species Goat
Immunogen NP_075555.
Antigen FOXL2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001277

 
novus biologicals canada
NB1001277
Unlabeled 0.1mg Each for $406.00

NB1001277SS

 
novus biologicals canada
NB1001277SS
Unlabeled 0.02mg Each for $142.00
Description & Specifications

Specifications

Host Species Goat
Immunogen NP_075555.
Antigen FOXL2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Peptide ELISA
Applications Western Blot
Test Specificity Expected from sequence similiarity: Rat, Cow .
Gene Accession No. P58012
Gene ID (Entrez) 668
Formulation Tris saline containing (pH 7.3) with 0.5% BSA
Concentration 0.5mg/mL