Please login to your online account to display your discounted pricing

anti-FOXL2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001277SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001277SS

  • / Each
 

Description & Specifications

Specifications

Antigen FOXL2
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Peptide ELISA
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation Tris saline containing (pH 7.3) with 0.5% BSA
Gene Accession No. P58012
Host Species Goat
Immunogen NP_075555.
Quantity 0.02mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 668
Test Specificity Expected from sequence similiarity: Rat, Cow .
Target Species Human
Target Species Murine