Please login to your online account to display your discounted pricing

anti-FoxC1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224834SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224834SS

  • / Each
 

Description & Specifications

Specifications

Antigen FoxC1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Antisera
Formulation Antibody in 200μL PBS with 0.2% gelatin
Gene Accession No. Q12948
Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human FOXC1 was used as the immunogen.
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2296
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, and 88% homologous in monkey.
Target Species Human
Target Species Primate