Please login to your online account to display your discounted pricing

anti-FoxC1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human FOXC1 was used as the immunogen.
Antigen FoxC1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224834

 
novus biologicals canada
NBP224834
Unlabeled 0.2mL Each for $255.80

NBP224834SS

 
novus biologicals canada
NBP224834SS
Unlabeled 0.05mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human FOXC1 was used as the immunogen.
Antigen FoxC1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, and 88% homologous in monkey.
Gene Accession No. Q12948
Gene ID (Entrez) 2296
Formulation Antibody in 200μL PBS with 0.2% gelatin