Please login to your online account to display your discounted pricing

anti-FNBP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224526

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224526

  • / Each
 

Description & Specifications

Specifications

Antigen FNBP1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q96RU3
Host Species Rabbit
Immunogen A portion of amino acids 30-80 of human FNBP1 was used as the immunogen for the antibody.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23048
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, orangutan, chimpanzee and rat, 94% homologous in cow and mouse, 88%homologous in dog, xenopus, tetradon, zebrafish and 83% homologous in opossum and chicken.
Target Species Human
Target Species Murine
Target Species Rat