Please login to your online account to display your discounted pricing

anti-FNBP1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 575-643 of human FNBP1 (isoform CRA_a) was used as the immunogen. 597 614
Antigen FNBP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227082SS

 
novus biologicals canada
NBP227082SS
Unlabeled 0.025mg Each for $139.00

NBP227082

 
novus biologicals canada
NBP227082
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 575-643 of human FNBP1 (isoform CRA_a) was used as the immunogen. 597 614
Antigen FNBP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_d), mouse, rat, chimp and horse.
Gene Accession No. Q96RU3
Gene ID (Entrez) 23048
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL