Please login to your online account to display your discounted pricing

anti-FNBP1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 30-80 of human FNBP1 was used as the immunogen for the antibody.
Antigen FNBP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224526SS

 
novus biologicals canada
NBP224526SS
Unlabeled 0.025mg Each for $139.00

NBP224526

 
novus biologicals canada
NBP224526
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 30-80 of human FNBP1 was used as the immunogen for the antibody.
Antigen FNBP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, orangutan, chimpanzee and rat, 94% homologous in cow and mouse, 88%homologous in dog, xenopus, tetradon, zebrafish and 83% homologous in opossum and chicken.
Gene Accession No. Q96RU3
Gene ID (Entrez) 23048
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL