Please login to your online account to display your discounted pricing

anti-FNBP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227082SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227082SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene Accession No. Q96RU3
Gene ID (Entrez) 23048
Immunogen A portion of amino acids 575-643 of human FNBP1 (isoform CRA_a) was used as the immunogen. 597 614
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_d), mouse, rat, chimp and horse.
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL
Antigen FNBP1
Host Species Rabbit
Regulatory Status RUO
Quantity 0.025mg
Applications Immunohistochemistry (Paraffin)
Format Purified