Please login to your online account to display your discounted pricing

anti-FNBP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227082

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227082

  • / Each
 

Description & Specifications

Specifications

Antigen FNBP1
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Gene Accession No. Q96RU3
Host Species Rabbit
Immunogen A portion of amino acids 575-643 of human FNBP1 (isoform CRA_a) was used as the immunogen. 597 614
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23048
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_d), mouse, rat, chimp and horse.
Target Species Equine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat