Please login to your online account to display your discounted pricing

anti-FMNL3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224724

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224724

  • / Each
 

Description & Specifications

Specifications

Antigen FMNL3
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q8IVF7
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human Formin-like protein 3 was used as the immunogen.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 91010
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_f), rhesus monkey and chimpanzee, and 72% homologous in cow.
Target Species Bovine
Target Species Canine
Target Species Equine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat