Please login to your online account to display your discounted pricing

anti-FMNL2, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 1-50 of human Formin-like protein 2 was used as the immunogen.
Antigen FMNL2
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224748SS

 
novus biologicals canada
NBP224748SS
Unlabeled 0.025mg Each for $139.00

NBP224748

 
novus biologicals canada
NBP224748
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 1-50 of human Formin-like protein 2 was used as the immunogen.
Antigen FMNL2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, monkey, rat and mouse, 94% homologous in cow, 92% homologous in chicken and 88% homologous in opossum.
Gene Accession No. Q9Y613
Gene ID (Entrez) 114793
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL