Please login to your online account to display your discounted pricing

anti-FMN2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223745

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223745

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 56776
Conjugate Unlabeled
Formulation 100μL of crude rabbit sera with 50% glycerol
Test Specificity This product is a formulation of IMG-80236, but is not the same exact product. The amino acid sequence used as immunogen is 100% homologous in human and mouse.
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Immunogen Within the range of amino acids 168-199 of human FMN2 protein were used as the immunogen.
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Antigen FMN2
Quantity 0.1mL
Format Antiserum
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal