Please login to your online account to display your discounted pricing

anti-FMN2, Polyclonal, Novus Biologicals™

$139.00 - $300.00

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 168-199 of human FMN2 protein were used as the immunogen.
Antigen FMN2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223745

 
novus biologicals canada
NBP223745
Unlabeled 0.1mL Each for $300.00

NBP223745SS

 
novus biologicals canada
NBP223745SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 168-199 of human FMN2 protein were used as the immunogen.
Antigen FMN2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity This product is a formulation of IMG-80236, but is not the same exact product. The amino acid sequence used as immunogen is 100% homologous in human and mouse.
Gene ID (Entrez) 56776
Formulation 100μL of crude rabbit sera with 50% glycerol