missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-FLJ10808, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human UBA6 was used as the immunogen for this antibody.
Antigen FLJ10808
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224587 Novus Biologicals Canada
NBP224587
0.1mg Each for $255.80
NBP224587SS Novus Biologicals Canada
NBP224587SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
FLJ10808
Affinity Purified
RUO
The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoforms CRA_a and CRA_b) and chimpanzee, and 93% homologous in monkey.
55236
0.5mg/mL
A portion of amino acids 300-350 of human UBA6 was used as the immunogen for this antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
A0AVT1
100μg antibody in 200μL PBS containing 0.05% BSA