Please login to your online account to display your discounted pricing

anti-FLJ10808, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 775-825 of human UBE1L2 was used as the immunogen for this antibody.
Antigen FLJ10808
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224534SS

 
novus biologicals canada
NBP224534SS
Unlabeled 0.025mg Each for $139.00

NBP224534

 
novus biologicals canada
NBP224534
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 775-825 of human UBE1L2 was used as the immunogen for this antibody.
Antigen FLJ10808
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoforms CRA_a and CRA_b) and 88% homologous in monkey.
Gene Accession No. A0AVT1
Gene ID (Entrez) 55236
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL