missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-FLIP, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A peptide corresponding to amino acids 206-222 GRSKEQRLKEQLGAQQE of human I-FLICE was used to immunize rabbits.
Antigen FLIP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056306 Novus Biologicals Canada
NB10056306
0.1mg Each for $255.80
NB10056306T Novus Biologicals Canada
NB10056306SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
FLIP
Affinity Purified
RUO
O15519
PBS with 0.05% BSA
A peptide corresponding to amino acids 206-222 GRSKEQRLKEQLGAQQE of human I-FLICE was used to immunize rabbits.
Polyclonal
Primary
Western Blot
8837
0.5mg/mL