Please login to your online account to display your discounted pricing

anti-Flightless I, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224584

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224584

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 2314
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Test Specificity The amino acid sequence used as immunogen for the hnRNPK antibody is 100% homologous in human, mouse, monkey, chimpanzee and rat, 94% homologous in dog and 78% homologous in chicken.
Concentration 0.5mg/mL
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Regulatory Status RUO
Antigen Flightless I
Monoclonal or Polyclonal Polyclonal
Immunogen A portion of amino acids 90-140 of human FLIIH was used as the immunogen for this antibody.
Format Affinity Purified
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Quantity 0.1mg