Please login to your online account to display your discounted pricing

anti-Flightless I, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 90-140 of human FLIIH was used as the immunogen for this antibody.
Antigen Flightless I
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224584SS

 
novus biologicals canada
NBP224584SS
Unlabeled 0.025mg Each for $109.00

NBP224584

 
novus biologicals canada
NBP224584
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 90-140 of human FLIIH was used as the immunogen for this antibody.
Antigen Flightless I
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen for the hnRNPK antibody is 100% homologous in human, mouse, monkey, chimpanzee and rat, 94% homologous in dog and 78% homologous in chicken.
Gene ID (Entrez) 2314
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL