Please login to your online account to display your discounted pricing

anti-Flightless I, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224584SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224584SS

  • / Each
 

Description & Specifications

Specifications

Antigen Flightless I
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 90-140 of human FLIIH was used as the immunogen for this antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2314
Test Specificity The amino acid sequence used as immunogen for the hnRNPK antibody is 100% homologous in human, mouse, monkey, chimpanzee and rat, 94% homologous in dog and 78% homologous in chicken.
Target Species Equine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat