Please login to your online account to display your discounted pricing

Fli-1 Polyclonal Antibody, Invitrogen™


Fli-1 Antibody, PA5-16899, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Western Blot.

Manufacturer: pierce biotechnology inc  PA516899

Catalog No. PIPA516899

  • / Each
 

Description & Specifications

Specifications

Applications Western Blot
Formulation 500μL purified antibody (0.2mg/mL) in PBS with 0.2% BSA and 15mM sodium azide; pH 7.6
Gene Alias Friend leukemia integration 1 transcription factor, proto-oncogene Fli-1, transcription factor ERGB
Gene ID (Entrez) 14247, 2313, 315532
Gene Accession No. P26323, Q01543
Isotype IgG
Gene Symbols FLI1
Species Reactivity Human, Mouse, Rat
Storage Requirements Store at 4° C
Host Species Rabbit
Immunogen Synthetic peptide derived from C-terminus of human Fli-1
Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Antigen Fli-1
Regulatory Status RUO
Quantity 500μL

Fli-1 Antibody, PA5-16899, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Western Blot.

Ets-1 is the prototype member of a family of genes identified on the basis of homology to the v-Ets oncogene isolated from the E26 erythroblastosis virus. This family of genes currently includes Ets-1, Ets-2, Erg-1, Erg-2, Elk, E74, Fli-1, PU.1 and PEA3. Members of the Ets gene family exhibit varied pattern of tissue expression, and share a highly conserved carboxy terminal domain containing a sequence related to the SV40 large T antigen nuclear localizartion signal sequence.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide derived from C-terminus of human Fli-1
Formulation: 500μL purified antibody (0.2mg/mL) in PBS with 0.2% BSA and 15mM sodium azide; pH 7.6
Synonyms: Friend leukemia integration 1 transcription factor, proto-oncogene Fli-1, transcription factor ERGB
Uniprot Accession: P26323, Q01543
Entrez GeneID: 14247, 2313, 315532
Gene Symbol: FLI1
PA5-16899 targets Fli-1 in WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-16899 immunogen is synthetic peptide derived from C-terminus of human Fli-1.