missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FHL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112859

Catalog No. 01-675-839



Description

Description

A suggested positive control for this product is Hela cell lysate.

A transcriptional coactivator, FHL2 is also known to form complexes consisting of Proline-, glutamic acid-, and leucine-rich protein-1 (PELP1), FHL2, and androgen receptors (AR) in prostate cancer cells, perhaps functioning as a molecular adaptor with PELP1 in the progression of prostate cancer.
Specifications

Specifications

FHL2
Polyclonal
PBS with 0.08% sodium azide
Q14192
DRAL, SLIM3
Rabbit
IgG
50 μg
-80° C, Avoid Freeze/Thaw Cycles
Primary
2274
Western Blot
Unconjugated
FHL2
Liquid
FHL2
Synthetic peptide corresponding to human FHL2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents