Please login to your online account to display your discounted pricing

anti-FHL2 Polyclonal, Thermo Scientific™

FHL2 Polyclonal antibody PA1-12870 from Thermo Scientific™ specifically detects FHL2 in Human samples, and it is validated for Western Blot

$471.25 - $651.05

Specifications

Antigen FHL2
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human FHL2.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-675-839

 
pierce biotechnology inc
PA112859
Unlabeled 50μg Each for $651.05

01-675-850

 
pierce biotechnology inc
PA112870
Unlabeled 100μg Each for $471.25
Description & Specifications

Specifications

Antigen FHL2
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human FHL2.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

FHL2 Polyclonal antibody PA1-12870 from Thermo Scientific™ specifically detects FHL2 in Human samples, and it is validated for Western Blot

A transcriptional coactivator, FHL2 is also known to form complexes consisting of Proline-, glutamic acid-, and leucine-rich protein-1 (PELP1), FHL2, and androgen receptors (AR) in prostate cancer cells, perhaps functioning as a molecular adaptor with PELP1 in the progression of prostate cancer.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic peptide corresponding to human FHL2.
Formulation: 100μg purified Antibody in PBS, with 0.08% sodium azide;
Synonyms: AAG11, DRAL, SLIM3
Uniprot Accession: Q14192
Gene Symbol: FHL2
Entrez GeneID: 2274
PA1-12870 detects FHL2 in Human samples. PA1-12870 has been successfully used in Western Blot procedures. PA1-12870 The immunogen is Synthetic peptide corresponding to human FHL2..