missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-FGF-20, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-171 of human FGF20 was used as the immunogen. 120 137
Antigen FGF-20
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227114 Novus Biologicals Canada
NBP227114
0.1mg Each for $257.41
NBP227114SS Novus Biologicals Canada
NBP227114SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
FGF-20
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat.
26281
0.5mg/mL
A portion of amino acids 100-171 of human FGF20 was used as the immunogen. 120 137
Polyclonal
Primary
Immunohistochemistry (Paraffin)
Q9NP95
100μg in 200μL PBS containing 0.05% BSA.